Assay Details
Target Gene Details
Entrez Gene ID: | 20014 |
Gene Name: | ribophorin II |
Gene Aliases: |
1300012C06Rik, AV261018, Rpn-2 |
Location: |
Chr.2:157279011-157326318 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpn2 | NM_019642.4 | NP_062616.2 | ||
| AK004968.1 | BAB23707.1 | |||
| AK018761.2 | ||||
| AK087923.1 | ||||
| AK153952.1 | BAE32275.1 | |||
| AK161624.1 | BAE36498.1 | |||
| AK166778.1 | BAE39013.1 | |||
| AK167864.1 | ||||
| AK167866.1 | ||||
| AK168221.1 | ||||
| BC010509.1 | AAH10509.1 | |||
| BC046806.1 | AAH46806.1 | |||
| D31717.1 | BAA06525.1 |
More Information
Set Membership: |
|

Genomic Map