Assay Details
Target Gene Details
Entrez Gene ID: | 111368 |
Gene Name: | prion protein gene complex |
Gene Aliases: |
Prnp-Prnd |
Location: |
Chr.2:131909928-131956131 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prn | NM_001278258.1 | NP_001265187.1 | ||
| NM_001278259.1 | NP_001265188.1 |
Target Gene Details
Entrez Gene ID: | 19122 |
Gene Name: | prion protein |
Gene Aliases: |
AA960666, AI325101, CD230, PrP, PrP<C>, PrPC, PrPSc, Prn-i, Prn-p, Sinc, prP27-30, prP33-35C |
Location: |
Chr.2:131909928-131938436 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prnp | NM_001278256.1 | NP_001265185.1 | ||
| NM_011170.3 | NP_035300.1 | |||
| AK148061.1 | BAE28320.1 | |||
| AK148892.1 | BAE28693.1 | |||
| AK150963.1 | BAE29994.1 | |||
| AK157840.1 | BAE34221.1 | |||
| AK158908.1 | BAE34724.1 | |||
| AK159056.1 | BAE34788.1 | |||
| AK159224.1 | BAE34911.1 | |||
| AK160091.1 | BAE35622.1 | |||
| AK162504.1 | BAE36950.1 | |||
| BC006703.1 | AAH06703.1 | |||
| CK622972.1 | ||||
| M13685.1 | AAA39996.1 | |||
| X79931.1 | 1 | 204 |

Genomic Map