Assay Details
Target Gene Details
Entrez Gene ID: | 64292 |
Gene Name: | prostaglandin E synthase |
Gene Aliases: |
2410099E23Rik, D2Ertd369e, Pges, mPGES, mPGES-1 |
Location: |
Chr.2:30889471-30903297 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptges | NM_022415.3 | 1 | 64 | NP_071860.1 |
| AB035323.1 | BAB71813.1 | |||
| AB041997.1 | BAA96083.1 | |||
| AK010757.1 | BAB27163.1 | |||
| AK033752.1 | BAC28463.1 | |||
| AK080897.1 | 1 | 69 | BAC38069.1 | |
| AK150013.1 | 1 | 57 | BAE29238.1 | |
| AK172621.1 | BAE43099.1 | |||
| BC024960.1 | AAH24960.1 |
More Information
Set Membership: |
|

Genomic Map