Assay Details
Target Gene Details
Entrez Gene ID: | 20614 |
Gene Name: | synaptosomal-associated protein 25 |
Gene Aliases: |
Bdr, GENA70, SNAP-25, SUP, sp |
Location: |
Chr.2:136713438-136782428 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snap25 | NM_001291056.1 | NP_001277985.1 | ||
| NM_011428.3 | NP_035558.1 | |||
| AF483516.1 | AAL90790.1 | |||
| AF483517.1 | AAL90791.1 | |||
| AK078038.1 | BAC37105.1 | |||
| AU080862.1 | ||||
| BC018249.1 | AAH18249.1 | |||
| CB232574.1 | ||||
| M22012.1 | AAA61741.1 |
More Information
Set Membership: |
|

Genomic Map