Assay Details
Target Gene Details
Entrez Gene ID: | 19223 |
Gene Name: | prostaglandin I2 (prostacyclin) synthase |
Gene Aliases: |
Cyp8, Cyp8a1, Pgis |
Location: |
Chr.2:167203194-167252127 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptgis | NM_008968.3 | 7 | 1102 | NP_032994.1 |
| AB001607.1 | 7 | 1020 | BAA21717.1 | |
| AK076585.1 | 7 | 1103 | BAC36403.1 | |
| AY285779.1 | 3 | 451 | AAP37445.1 | |
| BC062151.1 | 7 | 1109 | AAH62151.1 |

Genomic Map