Assay Details
Target Gene Details
Entrez Gene ID: | 19223 |
Gene Name: | prostaglandin I2 (prostacyclin) synthase |
Gene Aliases: |
Cyp8, Cyp8a1, Pgis |
Location: |
Chr.2:167203194-167252127 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptgis | NM_008968.3 | NP_032994.1 | ||
| AB001607.1 | BAA21717.1 | |||
| AK076585.1 | BAC36403.1 | |||
| BC062151.1 | AAH62151.1 |

Genomic Map