Assay Details
Target Gene Details
Entrez Gene ID: | 56045 |
Gene Name: | SAM domain and HD domain, 1 |
Gene Aliases: |
E330031J07Rik, Mg11 |
Location: |
Chr.2:157097529-157135277 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Samhd1 | NM_001139520.1 | NP_001132992.1 | ||
| NM_018851.3 | NP_061339.3 | |||
| AK054490.1 | BAC35801.1 | |||
| AK078936.1 | BAC37470.1 | |||
| AK128954.1 | BAC87661.1 | |||
| AK151335.1 | BAE30313.1 | |||
| AK153390.1 | BAE31954.1 | |||
| AK170688.1 | BAE41959.1 | |||
| AK171565.1 | ||||
| BC012721.1 | AAH12721.1 | |||
| BC067198.1 | AAH67198.1 | |||
| U15635.1 | AAA66219.1 |
More Information
Set Membership: |
|

Genomic Map