Assay Details
Target Gene Details
Entrez Gene ID: | 56045 |
Gene Name: | SAM domain and HD domain, 1 |
Gene Aliases: |
E330031J07Rik, Mg11 |
Location: |
Chr.2:157097529-157135277 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Samhd1 | NM_001139520.1 | 15 | 1840 | NP_001132992.1 |
| NM_018851.3 | 15 | 1840 | NP_061339.3 | |
| AK054490.1 | 15 | 1739 | BAC35801.1 | |
| AK128954.1 | 14 | 1638 | BAC87661.1 | |
| AK151335.1 | 15 | 1714 | BAE30313.1 | |
| AK153390.1 | 15 | 1714 | BAE31954.1 | |
| AK170688.1 | 15 | 1739 | BAE41959.1 | |
| AK171565.1 | 15 | 1740 | ||
| BC012721.1 | 15 | 1777 | AAH12721.1 | |
| BC067198.1 | 15 | 1763 | AAH67198.1 | |
| U15635.1 | 15 | 1764 | AAA66219.1 |
More Information
Set Membership: |
|

Genomic Map