Assay Details
Target Gene Details
Entrez Gene ID: | 227644 |
Gene Name: | small nuclear RNA activating complex, polypeptide 4 |
Gene Aliases: |
5730436L13Rik |
Location: |
Chr.2:26362765-26381072 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snapc4 | NM_001290419.1 | NP_001277348.1 | ||
| NM_172339.4 | NP_758842.1 | |||
| AK077522.1 | BAC36843.1 | |||
| BC044754.2 | AAH44754.1 | |||
| BC057031.1 | AAH57031.1 |

Genomic Map