Assay Details
Target Gene Details
Entrez Gene ID: | 66460 |
Gene Name: | SYS1 Golgi-localized integral membrane protein homolog (S. cerevisiae) |
Gene Aliases: |
2610042O14Rik, AI266964, C87389 |
Location: |
Chr.2:164456981-164479637 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sys1 | NM_025575.3 | NP_079851.2 | ||
| AK003071.1 | BAB22547.1 | |||
| AK005249.1 | ||||
| AK005510.1 | BAB24089.1 | |||
| AK007882.1 | ||||
| AK009684.1 | ||||
| AK011753.1 | ||||
| AK013220.1 | BAB28722.1 | |||
| AK017524.1 | ||||
| AK032073.1 | ||||
| AK032098.1 | ||||
| AK075770.1 | BAC35944.1 | |||
| AK148305.1 | BAE28468.1 | |||
| BC061084.1 | AAH61084.1 |
More Information
Set Membership: |
|

Genomic Map