Assay Details
Target Gene Details
Entrez Gene ID: | 19361 |
Gene Name: | RAD51 homolog |
Gene Aliases: |
AV304093, Rad51a, Reca |
Location: |
Chr.2:119112817-119136070 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rad51 | NM_011234.4 | NP_035364.1 | ||
| AK011242.1 | BAB27489.1 | |||
| AK045050.1 | ||||
| AK076468.1 | BAC36357.1 | |||
| AK151157.1 | BAE30162.1 | |||
| AK151177.1 | BAE30179.1 | |||
| AK160282.1 | ||||
| AK208342.1 | ||||
| BC027384.1 | AAH27384.1 | |||
| D13473.1 | BAA02718.1 | |||
| D13803.1 | BAA02961.1 |
More Information
Set Membership: |
|

Genomic Map