Assay Details
Target Gene Details
Entrez Gene ID: | 57314 |
Gene Name: | negative elongation factor complex member C/D, Th1l |
Gene Aliases: |
2410003I03Rik, C77797, Th1, Th1l |
Location: |
Chr.2:174415804-174427503 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nelfcd | NM_020580.3 | NP_065605.2 | ||
| AJ238741.1 | CAB65252.1 | |||
| AJ238742.1 | CAB65253.1 | |||
| AJ238743.1 | CAB65254.1 | |||
| AK010360.1 | BAB26880.1 | |||
| AK049418.1 | BAC33744.1 | |||
| AK159866.1 | BAE35440.1 | |||
| BC006959.1 | AAH06959.1 | |||
| BC031747.1 | AAH31747.1 |

Genomic Map