Assay Details
Target Gene Details
Entrez Gene ID: | 27223 |
Gene Name: | transformation related protein 53 binding protein 1 |
Gene Aliases: |
53BP1, Tp53bp1, m53BP1, p53BP1 |
Location: |
Chr.2:121194835-121290320 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trp53bp1 | NM_001290830.1 | 23 | 5193 | NP_001277759.1 |
| NM_013735.4 | 24 | 5343 | NP_038763.3 | |
| AJ414734.1 | 23 | 5186 | CAC94013.1 | |
| AK036324.1 | 5 | 1148 | BAC29383.1 | |
| AK132323.1 | 6 | 1376 | BAE21103.1 | |
| BC035206.1 | 12 | 2555 | AAH35206.1 | |
| BC079906.1 | 23 | 5127 | AAH79906.1 | |
| CA327869.1 |
More Information
Set Membership: |
|

Genomic Map