Assay Details
Target Gene Details
Entrez Gene ID: | 241514 |
Gene Name: | zinc finger protein 804A |
Gene Aliases: |
C630007C17Rik, Znf804a |
Location: |
Chr.2:82052874-82259879 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp804a | NM_175513.3 | NP_780722.2 | ||
AK043989.1 | BAC31727.2 | |||
AK049881.1 | BAC33969.1 |
