Assay Details
Target Gene Details
Entrez Gene ID: | 71481 |
Gene Name: | alpha-kinase 1 |
Gene Aliases: |
8430410J10Rik, LAK |
Location: |
Chr.3:127670308-127780553 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alpk1 | NM_027808.1 | NP_082084.1 | ||
| AK173191.1 | BAD32469.1 |
More Information
Set Membership: |
|

Genomic Map