Assay Details
Target Gene Details
Entrez Gene ID: | 27223 |
Gene Name: | transformation related protein 53 binding protein 1 |
Gene Aliases: |
53BP1, Tp53bp1, m53BP1, p53BP1 |
Location: |
Chr.2:121194835-121290320 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trp53bp1 | NM_001290830.1 | NP_001277759.1 | ||
| NM_013735.4 | NP_038763.3 | |||
| AJ414734.1 | CAC94013.1 | |||
| AK045851.1 | BAC32512.1 | |||
| AK148123.1 | BAE28360.1 | |||
| BC079906.1 | AAH79906.1 |
More Information
Set Membership: |
|

Genomic Map