Assay Details
Target Gene Details
Entrez Gene ID: | 72008 |
Gene Name: | zinc finger, FYVE domain containing 19 |
Gene Aliases: |
1500041L05Rik, ANCHR |
Location: |
Chr.2:119208720-119217050 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfyve19 | NM_001164827.1 | NP_001158299.1 | ||
| NM_028054.3 | NP_082330.2 | |||
| AK005386.1 | BAB23993.1 | |||
| BC018420.1 | AAH18420.1 | |||
| BC099429.1 | AAH99429.1 |

Genomic Map