Assay Details
Target Gene Details
Entrez Gene ID: | 75015 |
Gene Name: | RIKEN cDNA 4930503B20 gene |
Gene Aliases: |
1700013A07Rik, 1700095G21Rik, 4930483N21Rik, Gm1652, Samd13 |
Location: |
Chr.3:146645189-146686801 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930503B20Rik | NM_029144.3 | 2 | 896 | NP_083420.2 |
| AK005936.1 | 3 | 249 | ||
| AK015685.1 | 2 | 894 | ||
| BC048658.1 | 2 | 896 | AAH48658.1 |
Target Gene Details
Entrez Gene ID: | 100286843 |
Gene Name: | predicted gene 16325 |
Gene Aliases: |
OTTMUSG00000033652 |
Location: |
Chr.3:146641932-146651715 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm16325 | NR_045949.1 | |||
| AK139061.1 |
More Information
Set Membership: |
|

Genomic Map