Assay Details
Target Gene Details
Entrez Gene ID: | 74711 |
Gene Name: | tubulin tyrosine ligase-like family, member 9 |
Gene Aliases: |
1700016F23Rik, 4930509O20Rik, AW046572 |
Location: |
Chr.2:152950811-153008482 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttll9 | NM_001083618.1 | NP_001077087.1 | ||
| NM_029064.2 | NP_083340.2 | |||
| AK006027.1 | ||||
| AK015388.1 | ||||
| AK015740.1 | BAB29950.1 | |||
| AM690753.1 | ||||
| BC148285.2 |
More Information
Set Membership: |
|

Genomic Map