Assay Details
Target Gene Details
Entrez Gene ID: | 227613 |
Gene Name: | tubulin, beta 4B class IVB |
Gene Aliases: |
4930542G03Rik, Tubb2c, Tubb2c1 |
Location: |
Chr.2:25222158-25224702 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubb4b | NM_146116.2 | NP_666228.1 | ||
| AK015901.1 | ||||
| AK076895.1 | ||||
| AK146587.1 | BAE27282.1 | |||
| AK167022.1 | BAE39195.1 | |||
| BC005547.1 | AAH05547.1 | |||
| BC022919.1 | AAH22919.1 | |||
| BC083319.1 | AAH83319.1 |
More Information
Set Membership: |
|

Genomic Map