Assay Details
Target Gene Details
Entrez Gene ID: | 67187 |
Gene Name: | zinc finger, MYND domain containing 19 |
Gene Aliases: |
2700064H14Rik, AA536891, AU014915, mizip |
Location: |
Chr.2:24949802-24960200 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zmynd19 | NM_026021.3 | NP_080297.1 | ||
| AJ409150.1 | CAC84079.1 | |||
| AJ409151.1 | CAC84080.1 | |||
| AK005908.1 | BAB24312.1 | |||
| AK012484.1 | BAB28271.1 | |||
| AK088063.1 | ||||
| AK184476.1 | ||||
| BC027321.1 | AAH27321.1 |

Genomic Map