Assay Details
Target Gene Details
Entrez Gene ID: | 78635 |
Gene Name: | potassium voltage-gated channel, Shal-related family, member 3, opposite strand |
Gene Aliases: |
1700095B22Rik |
Location: |
Chr.3:105448190-105452955 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnd3os | NR_040759.1 | 2 | 481 | |
| AK131816.1 | 2 | 481 | BAE20817.1 |

Genomic Map