Assay Details
Target Gene Details
Entrez Gene ID: | 69126 |
Gene Name: | RIKEN cDNA 1810022K09 gene |
Gene Aliases: |
AI746582, AL023036 |
Location: |
Chr.3:14606289-14611256 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1810022K09Rik | NM_001099674.1 | NP_001093144.1 | ||
| AK007585.1 | ||||
| BC115807.1 |

Genomic Map