Assay Details
Target Gene Details
Entrez Gene ID: | 30931 |
Gene Name: | torsin family 1, member A (torsin A) |
Gene Aliases: |
DQ2, Dyt1, torsinA |
Location: |
Chr.2:30960561-30967918 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tor1a | NM_144884.2 | 1 | 189 | NP_659133.1 |
| AJ298841.1 | 1 | 144 | CAC12785.1 | |
| AK038891.1 | 1 | 145 | ||
| AK079728.1 | 1 | 143 | ||
| AK082936.1 | 1 | 99 | ||
| AK160361.1 | 1 | 145 | BAE35758.1 | |
| BC017683.1 | 1 | 169 | AAH17683.1 | |
| BY724297.1 | 1 | 178 |
More Information
Set Membership: |
|

Genomic Map