Assay Details
Target Gene Details
Entrez Gene ID: | 58203 |
Gene Name: | Z-DNA binding protein 1 |
Gene Aliases: |
2010010H03Rik, Dai, Dlm1, mZaDLM |
Location: |
Chr.2:173206612-173218922 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbp1 | NM_021394.2 | NP_067369.2 | ||
| AF136520.1 | AAF17234.1 | |||
| AK157215.1 | ||||
| AX775969.1 | CAE11684.1 | |||
| AX775971.1 | CAE11685.1 | |||
| BC020033.1 | AAH20033.1 |
More Information
Set Membership: |
|

Genomic Map