Assay Details
Target Gene Details
Entrez Gene ID: | 228880 |
Gene Name: | zinc finger, MYND-type containing 8 |
Gene Aliases: |
1110013E22Rik, 2010005I16Rik, 3632413B07Rik, AI316811, AL024039, Prkcbp1, RACK7 |
Location: |
Chr.2:165784152-165898984 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zmynd8 | NM_001252584.2 | 1 | 113 | NP_001239513.1 |
| NM_001252587.2 | 1 | 113 | NP_001239516.1 | |
| NM_027230.5 | 1 | 113 | NP_081506.3 | |
| BM933817.1 | ||||
| CN536818.1 | 1 | 148 |

Genomic Map