Assay Details
Target Gene Details
Entrez Gene ID: | 228880 |
Gene Name: | zinc finger, MYND-type containing 8 |
Gene Aliases: |
1110013E22Rik, 2010005I16Rik, 3632413B07Rik, AI316811, AL024039, Prkcbp1, RACK7 |
Location: |
Chr.2:165784152-165898984 on Build GRCm38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zmynd8 | NM_001252584.2 | NP_001239513.1 | ||
| NM_001252585.1 | NP_001239514.1 | |||
| NM_001252587.2 | NP_001239516.1 | |||
| NM_001281926.1 | NP_001268855.1 | |||
| NM_001291158.1 | NP_001278087.1 | |||
| NM_027230.5 | NP_081506.3 | |||
| AB093284.2 | BAC41468.4 | |||
| AK132427.1 | BAE21162.1 | |||
| AK147620.1 | BAE28029.1 | |||
| AK155864.1 | BAE33468.1 | |||
| BC048186.1 | AAH48186.3 | |||
| BC063267.1 | AAH63267.1 | |||
| CA328656.1 | ||||
| CK634740.1 | ||||
| CO428932.1 |
More Information
Set Membership: |
|

Genomic Map