Assay Details
Target Gene Details
Entrez Gene ID: | 99237 |
Gene Name: | transmembrane 9 superfamily protein member 4 |
Gene Aliases: |
AA986553, AU045326, B930079E06, mKIAA0255 |
Location: |
Chr.2:153161301-153210466 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tm9sf4 | NM_133847.3 | 14 | 1632 | NP_598608.2 |
| AB093220.2 | 13 | 1376 | BAC41404.3 | |
| AK035622.1 | 14 | 1594 | ||
| AK048158.1 | 14 | 1390 | BAC33261.1 | |
| AK083541.1 | 14 | 1632 | BAC38948.1 | |
| AK087048.1 | 14 | 1385 | BAC39789.1 | |
| AK088012.1 | 14 | 1388 | BAC40094.1 | |
| AK162435.1 | 14 | 1393 | BAE36916.1 | |
| AK171487.1 | 14 | 1390 | BAE42487.1 | |
| AK210804.1 | 5 | 355 | ||
| BC071208.1 | 1 | 451 | AAH71208.1 | |
| BC110309.1 | 14 | 1388 | AAI10310.1 | |
| BC116320.2 | 14 | 1584 |
More Information
Set Membership: |
|

Genomic Map