Assay Details
Target Gene Details
Entrez Gene ID: | 30934 |
Gene Name: | torsin family 1, member B |
Gene Aliases: |
2610016F05Rik, DQ1, torsinB |
Location: |
Chr.2:30952938-30959015 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tor1b | NM_133673.3 | 1 | 151 | NP_598434.2 |
| AJ297743.1 | 1 | 181 | CAC12814.1 | |
| AK017290.1 | 1 | 171 | ||
| AK028859.1 | ||||
| AK054404.1 | 1 | 181 | ||
| AK149754.1 | 1 | 172 | BAE29062.1 | |
| AK159843.1 | 1 | 150 | BAE35421.1 | |
| AK169692.1 | 1 | 174 | ||
| BC018456.1 | 1 | 150 | AAH18456.1 |
More Information
Set Membership: |
|

Genomic Map