Assay Details
Target Gene Details
Entrez Gene ID: | 69983 |
Gene Name: | sucrase isomaltase (alpha-glucosidase) |
Gene Aliases: |
2010204N08Rik, SI, Si-s |
Location: |
Chr.3:72888554-72967864 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 31 - Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sis | NM_001081137.1 | NP_001074606.1 | ||
| EU937530.1 |
More Information
Set Membership: |
|

Genomic Map