Assay Details
Target Gene Details
Entrez Gene ID: | 269389 |
Gene Name: | TOX high mobility group box family member 2 |
Gene Aliases: |
AI851523, AV026525, BI987407, Gcx1, RxHMG1 |
Location: |
Chr.2:163202643-163323102 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tox2 | NM_001098799.1 | NP_001092269.1 |

Genomic Map