Assay Details
Target Gene Details
Entrez Gene ID: | 66314 |
Gene Name: | tumor protein D52-like 2 |
Gene Aliases: |
2810411G23Rik, AU016537, C86069, D54 |
Location: |
Chr.2:181497104-181517962 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpd52l2 | NM_001291197.1 | 3 | 406 | NP_001278126.1 |
| NM_001291200.1 | 3 | 406 | NP_001278129.1 | |
| NM_001291201.1 | 3 | 406 | NP_001278130.1 | |
| NM_001291202.1 | 3 | 406 | NP_001278131.1 | |
| NM_001291203.1 | 3 | 406 | NP_001278132.1 | |
| NM_001291204.1 | NP_001278133.1 | |||
| NM_025482.3 | 3 | 406 | NP_079758.2 | |
| NR_111899.1 | 2 | 260 | ||
| AA218395.1 | ||||
| AK013085.1 | 3 | 348 | BAB28639.1 | |
| AK013188.1 | 3 | 359 | BAB28701.1 | |
| AK051268.1 | 3 | 350 | BAC34583.1 | |
| AK147885.1 | 3 | 366 | BAE28203.1 | |
| AK155987.1 | 3 | 343 | BAE33536.1 | |
| AK159925.1 | 3 | 334 | BAE35487.1 | |
| AK160727.1 | 3 | 350 | BAE35972.1 | |
| AK161092.1 | 3 | 362 | BAE36191.1 | |
| AK171825.1 | 3 | 368 | BAE42684.1 | |
| AK199788.1 | 1 | 65 | ||
| BC006886.1 | 3 | 295 | AAH06886.1 | |
| CN721218.1 | 2 | 260 | ||
| DV047279.1 | 3 | 392 |

Genomic Map