Assay Details
Target Gene Details
Entrez Gene ID: | 70296 |
Gene Name: | TBC1 domain family, member 13 |
Gene Aliases: |
2600014A06Rik, BC025586 |
Location: |
Chr.2:30133751-30152013 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d13 | NM_146252.2 | NP_666364.1 | ||
| AK158066.1 | BAE34347.1 | |||
| BC025586.1 | AAH25586.1 |
More Information
Set Membership: |
|

Genomic Map