Assay Details
Target Gene Details
Entrez Gene ID: | 67231 |
Gene Name: | TBC1 domain family, member 20 |
Gene Aliases: |
1110028I04Rik, 2810442O16Rik, AI414693 |
Location: |
Chr.2:152293872-152312590 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d20 | NM_024196.3 | 4 | 493 | NP_077158.1 |
| AK006877.1 | 4 | 490 | BAB24777.1 | |
| AK076124.1 | 4 | 494 | BAC36202.1 | |
| BC002196.1 | 3 | 309 | AAH02196.1 | |
| BC040089.1 | 4 | 456 | AAH40089.1 |
More Information
Set Membership: |
|

Genomic Map