Assay Details
Target Gene Details
Entrez Gene ID: | 67231 |
Gene Name: | TBC1 domain family, member 20 |
Gene Aliases: |
1110028I04Rik, 2810442O16Rik, AI414693 |
Location: |
Chr.2:152293872-152312590 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d20 | NM_024196.3 | 7 | 923 | NP_077158.1 |
| AK006877.1 | 7 | 920 | BAB24777.1 | |
| AK076124.1 | 7 | 924 | BAC36202.1 | |
| AK177685.1 | 2 | 123 | ||
| AK198420.1 | ||||
| BC002196.1 | 6 | 739 | AAH02196.1 | |
| BC040089.1 | 7 | 886 | AAH40089.1 |
More Information
Set Membership: |
|

Genomic Map