Assay Details
Target Gene Details
Entrez Gene ID: | 228801 |
Gene Name: | BPI fold containing family B, member 1 |
Gene Aliases: |
Lplunc1 |
Location: |
Chr.2:154190818-154220343 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bpifb1 | NM_001012392.1 | NP_001012392.1 | ||
| NM_153418.2 | NP_700467.2 | |||
| AK146605.1 | ||||
| BC060982.1 | AAH60982.1 | |||
| BC100436.1 | AAI00437.1 | |||
| U46068.3 | AAA87581.3 |

Genomic Map