Assay Details
Target Gene Details
Entrez Gene ID: | 71861 |
Gene Name: | zinc finger SWIM-type containing 2 |
Gene Aliases: |
1700025P14Rik, 4933437F18Rik, MEX |
Location: |
Chr.2:83915079-83941254 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zswim2 | NM_027964.2 | NP_082240.1 | ||
| AK006367.1 | BAB24549.1 | |||
| AK017097.1 | ||||
| BC120837.1 | ||||
| EU233996.1 |
More Information
Set Membership: |
|

Genomic Map