Assay Details
Target Gene Details
Entrez Gene ID: | 98766 |
Gene Name: | ubiquitin associated domain containing 1 |
Gene Aliases: |
1110033G07Rik, AA407978, GBDR1, Gdbr1, Ubadc1 |
Location: |
Chr.2:25996958-26021781 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ubac1 | NM_133835.2 | NP_598596.2 | ||
| AK157857.1 | BAE34235.1 | |||
| AK206541.1 | ||||
| BC021811.1 | AAH21811.1 |

Genomic Map