Assay Details
Target Gene Details
Entrez Gene ID: | 68612 |
Gene Name: | ubiquitin-conjugating enzyme E2C |
Gene Aliases: |
1110015A16Rik, D2Ertd695e |
Location: |
Chr.2:164769929-164772902 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2c | NM_026785.2 | NP_081061.1 | ||
| AK003722.1 | BAB22959.1 | |||
| AK136186.1 | BAE22863.1 | |||
| AK160740.1 | BAE35981.1 | |||
| BC147032.1 | ||||
| BC147033.1 |
More Information
Set Membership: |
|

Genomic Map