Assay Details
Target Gene Details
Entrez Gene ID: | 11608 |
Gene Name: | angiotensin II receptor, type 1b |
Gene Aliases: |
AT1B, AT2R1B, Agtr-1b, Angtr-1b |
Location: |
Chr.3:20314473-20367177 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agtr1b | NM_175086.3 | 1 | 76 | NP_780295.2 |
| AK087228.1 | 1 | 76 | BAC39824.1 | |
| BC109153.1 | AAI09154.1 |
More Information
Set Membership: |
|

Genomic Map