Assay Details
Target Gene Details
Entrez Gene ID: | 22029 |
Gene Name: | TNF receptor-associated factor 1 |
Gene Aliases: |
4732496E14Rik |
Location: |
Chr.2:34943256-34961772 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Traf1 | NM_009421.3 | NP_033447.2 | ||
| AK029135.1 | BAC26315.1 | |||
| AK089281.1 | BAC40826.1 | |||
| AK142931.1 | BAE25226.1 | |||
| AK157375.1 | ||||
| AK169515.1 | BAE41205.1 | |||
| L35302.1 | AAC37663.1 |
More Information
Set Membership: |
|

Genomic Map