Assay Details
Target Gene Details
Entrez Gene ID: | 22029 |
Gene Name: | TNF receptor-associated factor 1 |
Gene Aliases: |
4732496E14Rik |
Location: |
Chr.2:34943256-34961772 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Traf1 | NM_009421.3 | 9 | 1298 | NP_033447.2 |
| AK029135.1 | 9 | 1300 | BAC26315.1 | |
| AK169515.1 | 9 | 1091 | BAE41205.1 | |
| AK172598.1 | 5 | 902 | BAE43086.1 | |
| BC096595.1 | 5 | 864 | AAH96595.1 | |
| L35302.1 | 8 | 1044 | AAC37663.1 |
More Information
Set Membership: |
|

Genomic Map