Assay Details
Target Gene Details
Entrez Gene ID: | 215280 |
Gene Name: | WAS/WASL interacting protein family, member 1 |
Gene Aliases: |
AI115543, D2Ertd120e, WIP, Waspip |
Location: |
Chr.2:73429610-73529733 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wipf1 | NM_001289722.1 | 6 | 1411 | NP_001276651.1 |
| NM_001289723.1 | 6 | 1376 | NP_001276652.1 | |
| NM_153138.4 | 6 | 1406 | NP_694778.1 | |
| AJ437262.1 | 5 | 1207 | CAD24773.1 | |
| AK031046.1 | 6 | 1373 | BAC27228.1 | |
| AK156547.1 | 6 | 1411 | BAE33753.1 | |
| AK168092.1 | 2 | 560 | BAE40065.1 | |
| BC049788.1 | 6 | 1329 | AAH49788.1 |
More Information
Set Membership: |
|

Genomic Map