Assay Details
Target Gene Details
Entrez Gene ID: | 228775 |
Gene Name: | tribbles homolog 3 (Drosophila) |
Gene Aliases: |
Ifld2, Nipk, SINK, SKIP3, TRB-3, Trb3 |
Location: |
Chr.2:152337425-152344060 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trib3 | NM_175093.2 | 1 | 105 | NP_780302.2 |
| AF358868.1 | 1 | 82 | AAM45476.1 | |
| AJ514260.1 | CAD55728.1 | |||
| AK089931.1 | BAC41002.1 | |||
| AK132257.1 | BAE21062.1 | |||
| AK144752.1 | BAE26048.1 | |||
| AK146340.1 | BAE27094.1 | |||
| AK159760.1 | BAE35351.1 | |||
| AK160099.1 | 1 | 96 | ||
| BC012955.1 | 1 | 77 | AAH12955.1 | |
| BY010029.1 | 1 | 106 |
More Information
Set Membership: |
|

Genomic Map