Assay Details
Target Gene Details
Entrez Gene ID: | 241431 |
Gene Name: | xin actin-binding repeat containing 2 |
Gene Aliases: |
2310003D02Rik, 2310008C07Rik, A530024P18Rik, AI452089, Cmya3, Gm352, Xin2 |
Location: |
Chr.2:67446000-67526620 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xirp2 | NM_001024618.2 | NP_001019789.1 | ||
| NM_001083919.1 | NP_001077388.1 | |||
| AK040786.1 | BAC30703.1 | |||
| AY775570.1 | AAV52766.1 | |||
| DQ011666.1 | AAY44537.1 | |||
| EF119712.1 | ||||
| EF119716.1 | ||||
| EF122140.1 | ||||
| EU286531.1 |
More Information
Set Membership: |
|

Genomic Map