Assay Details
Target Gene Details
Entrez Gene ID: | 68521 |
Gene Name: | family with sequence similarity 189, member B |
Gene Aliases: |
1110013L07Rik |
Location: |
Chr.3:89183142-89189291 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam189b | NM_001014995.1 | 6 | 1248 | NP_001014995.1 |
| BC088990.1 | 6 | 1248 | AAH88990.1 |

Genomic Map