Assay Details
Target Gene Details
Entrez Gene ID: | 68659 |
Gene Name: | family with sequence similarity 198, member B |
Gene Aliases: |
1110032E23Rik, 2210419I08Rik, AV011458, Ened |
Location: |
Chr.3:79884475-79946278 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam198b | NM_133187.3 | 2 | 1340 | NP_573450.2 |
| AF285091.1 | 2 | 1282 | AAG00502.1 | |
| AK052367.1 | 2 | 307 | BAC34959.1 | |
| AK143522.1 | 2 | 1071 | BAE25414.1 | |
| BC056975.1 | 2 | 1109 | AAH56975.1 | |
| EU797522.1 | 2 | 1118 |

Genomic Map