Assay Details
Target Gene Details
Entrez Gene ID: | 71643 |
Gene Name: | zinc finger, GRF-type containing 1 |
Gene Aliases: |
4930422G04Rik, AI448607 |
Location: |
Chr.3:127553436-127618023 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zgrf1 | NM_197997.2 | 5 | 450 | NP_932114.2 |
| AY512952.1 | 3 | 2943 | AAR87822.1 | |
| BC085187.1 | 5 | 306 |

Genomic Map