Assay Details
Target Gene Details
Entrez Gene ID: | 76295 |
Gene Name: | ATPase, class VI, type 11B |
Gene Aliases: |
1110019I14Rik, ATPIF, ATPIR, mKIAA0956 |
Location: |
Chr.3:35754062-35856276 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp11b | NM_029570.3 | 7 | 845 | NP_083846.2 |
| AK129248.1 | 7 | 837 | BAC98058.1 | |
| AK154156.1 | 7 | 849 | ||
| AK155372.1 | 7 | 849 | BAE33225.1 | |
| BC076603.1 | 7 | 715 | AAH76603.1 |
More Information
Set Membership: |
|

Genomic Map