Assay Details
Target Gene Details
Entrez Gene ID: | 12904 |
Gene Name: | cellular retinoic acid binding protein II |
Gene Aliases: |
AI893628, Crabp-2, CrabpII |
Location: |
Chr.3:87948693-87953372 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crabp2 | NM_007759.2 | NP_031785.1 | ||
| BC018397.1 | AAH18397.1 | |||
| M35523.1 | AAA37454.1 |

Genomic Map